Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025641.3 | NP_079917.1 | 3 | 248 | ||
GenBank mRNA | AK003266.1 | - | 3 | 213 | ||
AK003539.1 | - | 3 | 191 | |||
AK011734.1 | - | 610 | ||||
AK012552.1 | - | 3 | 226 | |||
AK018712.1 | - | 3 | 226 | |||
AK054005.1 | - | 3 | 214 | |||
AK088185.1 | - | 3 | 213 | |||
BC011388.1 | - | 3 | 192 | 3597862 | ||
BY138998.1 | - | 3 | 248 | |||
BY702808.1 | - | 3 | 221 |