Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159609.1 | NP_001153081.1 | 3 | 320 | ||
NM_001159610.1 | NP_001153082.1 | 3 | 371 | |||
NM_001159612.1 | NP_001153084.1 | 3 | 387 | |||
NM_025657.3 | NP_079933.2 | 3 | 270 | |||
GenBank mRNA | AK003037.1 | - | 3 | 177 | ||
AK003592.1 | - | 3 | 288 | |||
AK012637.1 | - | 3 | 385 | |||
AK033084.1 | - | 1 | 582 | |||
AK134971.1 | - | 3 | 229 | |||
AK169249.1 | - | 3 | 294 | |||
BC034894.1 | - | 3 | 229 | 3328402 | ||
BU604191.1 | - | 3 | 273 | |||
BY293050.1 | - | 3 | 246 | |||
CJ169080.1 | - | 3 | 389 |