Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006636.3 | NP_006627.2 | 4 | 543 | ||
NR_027405.1 | 5 | 641 | ||||
XM_006711924.2 | XP_006711987.1 | 3 | 330 | |||
GenBank mRNA | AK130664.1 | - | 4 | 491 | ||
AK222826.1 | - | 4 | 493 | |||
AK223041.1 | - | 4 | 491 | |||
AK300035.1 | - | 3 | 308 | |||
AK302245.1 | - | 2 | 183 | |||
AK313138.1 | - | 4 | 491 | |||
AU125762.1 | - | 4 | 522 | |||
BC001548.1 | - | 4 | 478 | 3462450 | ||
BC015062.1 | - | 5 | 591 | 4285669 | ||
BC017054.1 | - | 4 | 471 | 3846710 | ||
BX475613.1 | - | 4 | 544 | |||
X16396.1 | - | 4 | 461 |