Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110809.2 | NP_001104279.1 | 8 | 1136 | ||
NM_001286857.1 | NP_001273786.1 | 8 | 1750 | |||
NM_025663.4 | NP_079939.3 | 9 | 1271 | |||
GenBank mRNA | AK011617.1 | - | 8 | 1134 | ||
AK018306.1 | - | 8 | 1110 | |||
AK169111.1 | - | 8 | 1125 | |||
BC019490.1 | - | 10 | 992 | 4039649 | ||
BC080676.1 | - | 8 | 1728 |