Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025664.5 | NP_079940.2 | 8 | 941 | ||
GenBank mRNA | AK012520.1 | - | 3 | 233 | ||
AK139802.1 | - | 8 | 975 | |||
AK149739.1 | - | 8 | 955 | |||
AK149849.1 | - | 8 | 995 | |||
AK155830.1 | - | 976 | ||||
AK157517.1 | - | 8 | 957 | |||
AK170339.1 | - | 957 | ||||
BC014814.1 | - | 8 | 941 | 3257422 |