Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025666.2 | NP_079942.1 | 2 | 316 | ||
GenBank mRNA | AK011968.1 | - | 2 | 187 | ||
AK017542.1 | - | 2 | 271 | |||
AK088208.1 | - | 2 | 318 | |||
AK146308.1 | - | 2 | 317 | |||
AK149989.1 | - | 2 | 271 | |||
AK155611.1 | - | 2 | 187 | |||
AK190790.1 | - | 1 | 50 | |||
BC079531.1 | - | 1 | 13 | |||
BC094323.1 | - | 2 | 324 | |||
BC094423.1 | - | 2 | 274 | 6812389 |