Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025670.4 | NP_079946.2 | 5 | 469 | ||
GenBank mRNA | AK005157.1 | - | 5 | 455 | ||
AK017480.1 | - | 5 | 452 | |||
AK031705.1 | - | 5 | 471 | |||
AK035791.1 | - | 3 | 1764 | |||
AK051560.1 | - | 5 | 467 | |||
AK149939.1 | - | 5 | 465 | |||
AK166141.1 | - | 5 | 462 | |||
BC004063.1 | - | 5 | 468 | 3590511 |