Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lix1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025681.2 | NP_079957.2 | 5, 6 | 828 | ||
GenBank mRNA | AK012220.1 | - | 5, 6 | 636 | ||
AK017685.1 | - | 5, 6 | 647 | |||
AK161930.1 | - | 2, 3 | 405 | |||
BC049574.1 | - | 5, 6 | 842 | 6593108 | ||
BC052700.1 | - | 5, 6 | 628 | 6416835 | ||
BC063057.1 | - | 5, 6 | 621 | 5715784 |