Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCCAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039707.1 | NP_001034796.1 | 6 | 1074 | ||
NM_001039708.1 | NP_001034797.1 | 4 | 855 | |||
NM_006643.3 | NP_006634.3 | 5 | 1005 | |||
XM_005266050.3 | XP_005266107.1 | 6 | 1028 | |||
XM_005266051.3 | XP_005266108.1 | 5 | 919 | |||
XM_011518157.2 | XP_011516459.1 | 5 | 858 | |||
XM_017014218.1 | XP_016869707.1 | 5 | 1175 | |||
XM_017014219.1 | XP_016869708.1 | 5 | 1175 | |||
GenBank mRNA | AF039688.1 | - | 2 | 142 | ||
AK001296.1 | - | 4 | 634 | |||
AY349357.1 | - | 5 | 695 | |||
BC014515.1 | - | 5 | 872 | 3547130 |