Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025683.2 | NP_079959.2 | 5 | 516 | ||
GenBank mRNA | AK007735.1 | - | 5 | 522 | ||
AK017774.1 | - | 5 | 517 | |||
AK029088.1 | - | 5 | 515 | |||
AK088152.1 | - | 5 | 517 | |||
AK150008.1 | - | 5 | 554 | |||
BC006953.1 | - | 5 | 510 | |||
BC019126.1 | - | 5 | 559 | 5008075 |