Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025683.2 | NP_079959.2 | 6 | 1261 | ||
GenBank mRNA | AK007735.1 | - | 6 | 1267 | ||
AK017774.1 | - | 6 | 1262 | |||
AK029088.1 | - | 6 | 1260 | |||
AK088152.1 | - | 6 | 1262 | |||
AK150008.1 | - | 6 | 1300 | |||
BC006953.1 | - | 6 | 1255 |