Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025686.2 | NP_079962.1 | 2, 3 | 277 | ||
GenBank mRNA | AK012459.1 | - | 1, 2 | 1179 | ||
AK012680.1 | - | 2, 3 | 278 | |||
AK017767.1 | - | 2, 3 | 280 | |||
AK028228.1 | - | 2, 3 | 278 | |||
AK083122.1 | - | 2, 3 | 278 | |||
AK151198.1 | - | 2, 3 | 278 | |||
AK152842.1 | - | 2, 3 | 297 | |||
BC021918.1 | - | 264 | ||||
BC098232.1 | - | 2, 3 | 258 | 2938123 |