Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025687.3 | NP_079963.1 | 3, 4 | 227 | ||
GenBank mRNA | AF285582.1 | - | 3, 4 | 285 | ||
AK006285.1 | - | 3, 4 | 271 | |||
AK014896.1 | - | 3, 4 | 284 | |||
AK017776.1 | - | 3, 4 | 283 | |||
BC048458.1 | - | 3, 4 | 227 | 6775116 | ||
BC061081.1 | - | 3, 4 | 282 | 6773183 |