Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025689.4 | NP_079965.2 | 4 | 871 | ||
GenBank mRNA | AK014056.1 | - | 3 | 811 | ||
AK017860.1 | - | 4 | 865 | |||
AK137013.1 | - | 4 | 875 | |||
AK141240.1 | - | 4 | 874 | |||
BC098221.1 | - | 3 | 817 | 949536 | ||
BC116786.1 | - | 3 | 729 | 40126105 | ||
BC116788.1 | - | 3 | 729 | 40126107 |