Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkapl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025719.3 | NP_079995.2 | 1 | 592 | ||
GenBank mRNA | AK014812.1 | - | 1 | 579 | ||
AK015912.1 | - | 1 | 574 | |||
AK160424.1 | - | 1 | 394 | |||
BC048457.1 | - | 1 | 578 | |||
BC061080.1 | - | 1 | 584 | |||
BC089537.1 | - | 1 | 580 | |||
BC099550.1 | - | 1 | 592 | 30918538 |