Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Henmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078646.1 | NP_001072114.1 | 6 | 708 | ||
NM_025723.2 | NP_079999.2 | 6 | 1023 | |||
GenBank mRNA | AK014900.1 | - | 6 | 989 | ||
AK038994.1 | - | 6 | 709 | |||
AK085587.1 | - | 6 | 1023 | |||
BC099408.1 | - | 5 | 689 | 30935249 | ||
BC116412.1 | - | 4 | 382 | |||
BC116413.1 | - | 4 | 504 | |||
BC128494.1 | - | 4 | 504 | |||
BC128495.1 | - | 4 | 504 |