Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933400A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015979.1 | - | 2 | 336 | ||
AK016585.1 | - | 3 | 470 | |||
BC107240.1 | - | 3 | 387 | |||
BC107241.1 | - | 3 | 391 |