Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gid4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025757.4 | NP_080033.3 | 6 | 1886 | ||
GenBank mRNA | AJ404329.1 | - | 6 | 1380 | ||
AK039435.1 | - | 6 | 1770 | |||
AK077759.1 | - | 6 | 1454 | |||
AK164644.1 | - | 6 | 1753 | |||
BC044901.1 | - | 6 | 1588 | 3660276 | ||
BC046821.1 | - | 6 | 1700 | 6400140 | ||
BC052418.1 | - | 6 | 1728 | 6401069 |