Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUVBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321190.1 | NP_001308119.1 | 6 | 941 | ||
NM_001321191.1 | NP_001308120.1 | 6 | 850 | |||
NM_006666.2 | NP_006657.1 | 6 | 871 | |||
NR_135578.1 | 6 | 871 | ||||
XM_011526330.1 | XP_011524632.1 | 6 | 784 | |||
GenBank mRNA | AB024301.1 | - | 6 | 420 | ||
AF124607.1 | - | 6 | 428 | |||
AF151804.1 | - | 6 | 413 | |||
AF155138.1 | - | 6 | 413 | |||
AK027762.1 | - | 6 | 491 | |||
AK057498.1 | - | 6 | 1418 | |||
AK074542.1 | - | 6 | 421 | |||
AK301344.1 | - | 6 | 429 | |||
AL136743.1 | - | 5 | 410 | |||
BC000428.2 | - | 6 | 412 | 2819778 | ||
BC004531.2 | - | 6 | 420 | 3051517 | ||
BC008355.1 | - | 6 | 420 | |||
CR533507.1 | - | 5 | 407 | |||
Y18417.1 | - | 6 | 412 |