Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025768.2 | NP_080044.2 | 8 | 1225 | ||
GenBank mRNA | AF329833.1 | - | 6 | 1013 | ||
AK007905.1 | - | 8 | 1230 | |||
AK017197.1 | - | 8 | 1189 | |||
AK017251.1 | - | 9 | 1187 | |||
AK149322.1 | - | 1222 | ||||
BC029096.1 | - | 8 | 1231 | 3500563 |