Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025769.3 | NP_080045.1 | 4 | 553 | ||
GenBank mRNA | AK015426.1 | - | 535 | |||
AK015866.1 | - | 4 | 573 | |||
AK017267.1 | - | 4 | 554 | |||
AK030381.1 | - | 4 | 563 | |||
AK038735.1 | - | 4 | 563 | |||
AK042748.1 | - | 4 | 548 | |||
AK043519.1 | - | 4 | 525 | |||
AK077722.1 | - | 4 | 556 | |||
BC085167.1 | - | 4 | 445 | 30538702 |