Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025770.3 | NP_080046.3 | 4 | 281 | ||
GenBank mRNA | AB079383.1 | - | 3 | 237 | ||
AB091691.1 | - | 3 | 258 | |||
AK009076.1 | - | 4 | 265 | |||
AK017355.1 | - | 4 | 263 | |||
AK077333.1 | - | 4 | 254 | |||
BB843097.1 | - | 4 | 281 | |||
BC023455.1 | - | 3 | 257 | 5249268 |