Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025770.3 | NP_080046.3 | 8 | 920 | ||
GenBank mRNA | AB091691.1 | - | 7 | 897 | ||
AK009076.1 | - | 8 | 904 | |||
AK017355.1 | - | 8 | 902 | |||
AK077333.1 | - | 8 | 893 | |||
BC023455.1 | - | 7 | 896 | 5249268 |