Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Duoxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025777.2 | NP_080053.1 | 6 | 1032 | ||
GenBank mRNA | AK008816.1 | - | 6 | 1032 | ||
AK018569.1 | - | 6 | 1034 | |||
BC031111.1 | - | 6 | 1019 | 4989815 |