Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025780.3 | NP_080056.1 | 3 | 1154 | ||
GenBank mRNA | AK009162.1 | - | 3 | 1135 | ||
AK018577.1 | - | 3 | 1134 | |||
AK051198.1 | - | 2 | 934 | |||
AK081303.1 | - | 3 | 1155 | |||
BC027502.1 | - | 3 | 1062 | 1515752 |