Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nacc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025788.3 | NP_080064.3 | 6 | 4036 | ||
GenBank mRNA | AK008006.1 | - | 1 | 511 | ||
BC048157.1 | - | 6 | 4009 | 5697941 | ||
BC052706.1 | - | 6 | 4019 | 5702150 | ||
BC054800.1 | - | 6 | 4036 | 6850337 |