Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025789.5 | NP_080065.4 | 8 | 1556 | ||
GenBank mRNA | AK007979.1 | - | 8 | 1189 | ||
AK016364.1 | - | 8 | 1142 | |||
AK148230.1 | - | 8 | 1518 | |||
BC054439.1 | - | 8 | 1224 | |||
BC057649.1 | - | 8 | 1515 | |||
BC085294.1 | - | 8 | 1204 | |||
BC150663.1 | - | 8 | 1041 | 9087574 |
Gene Symbol: Rsph3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083945.1 | NP_001077414.1 | 8 | 1245 | ||
GenBank mRNA | AK006083.1 | - | 8 | 1245 | ||
AK016473.1 | - | 8 | 1203 | |||
AK076872.1 | - | 8 | 1247 | |||
AK161285.1 | - | 8 | 1208 | |||
BC110284.1 | - | 8 | 1531 | 5150198 |