Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025796.3 | NP_080072.2 | 4 | 412 | ||
GenBank mRNA | AB049651.1 | - | 4 | 395 | ||
AK002421.1 | - | 4 | 392 | |||
AK008431.1 | - | 4 | 392 | |||
AK021173.1 | - | 4 | 392 | |||
AK041771.1 | - | 2 | 2690 | |||
AK167883.1 | - | 4 | 392 | |||
BC006029.1 | - | 2 | 2676 | |||
BC037363.1 | - | 4 | 378 | 1514355 | ||
BC051471.1 | - | 4 | 389 | 1514101 | ||
BC055724.1 | - | 4 | 371 | 6411227 | ||
BC059722.1 | - | 4 | 391 | 6772871 |