Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163689.1 | NP_001157161.1 | 9 | 1751 | ||
NM_025802.3 | NP_080078.2 | 8 | 1583 | |||
GenBank mRNA | AK002826.1 | - | 8 | 1565 | ||
AK003207.1 | - | 4 | 806 | |||
AK031609.1 | - | 9 | 1709 | |||
AK076464.1 | - | 9 | 1536 | |||
AK085926.1 | - | 9 | 1736 | |||
AK150184.1 | - | 9 | 1733 | |||
AK185834.1 | - | 1 | 94 | |||
AY510273.1 | - | 9 | 1732 | |||
AY731699.1 | - | 9 | 1732 | |||
BC019188.1 | - | 10 | 1815 | 5027544 | ||
BC026807.1 | - | 1 | 36 | |||
BC034190.1 | - | 8 | 1252 | |||
BC044737.1 | - | 10 | 1797 | |||
BC044781.2 | - | 10 | 1797 | 5151911 | ||
BC054825.1 | - | 10 | 1713 | |||
BC064747.1 | - | 9 | 1723 | 30024535 |