Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006678.4 | NP_006669.1 | 4 | 1468 | ||
GenBank mRNA | BC022279.1 | - | 4 | 1440 | 4692025 |