Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ME3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014811.1 | NP_001014811.1 | 6 | 1024 | ||
NM_001161586.1 | NP_001155058.1 | 7 | 1097 | |||
NM_006680.2 | NP_006671.2 | 7 | 872 | |||
XM_005273717.1 | XP_005273774.1 | 7 | 973 | |||
XM_017017136.1 | XP_016872625.1 | 7 | 1855 | |||
XM_017017137.1 | XP_016872626.1 | 3 | 397 | |||
XM_017017138.1 | XP_016872627.1 | 8 | 981 | |||
GenBank mRNA | AA403046.1 | - | 2 | 112 | ||
AA449933.1 | - | 2 | 112 | |||
AK092181.1 | - | 7 | 835 | |||
AK300974.1 | - | 7 | 1097 | |||
AK313695.1 | - | 7 | 947 | |||
AY424278.1 | - | 5 | 584 | |||
BC022472.2 | - | 6 | 1024 | 4812045 | ||
X79440.1 | - | 6 | 802 |