Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025813.3 | NP_080089.1 | 3 | 291 | ||
GenBank mRNA | AK004586.1 | - | 3 | 292 | ||
AK032688.1 | - | 3 | 260 | |||
AK075859.1 | - | 3 | 321 | |||
AK135235.1 | - | 3 | 292 | |||
AK136108.1 | - | 3 | 283 | |||
AK152444.1 | - | 3 | 313 | |||
AK159554.1 | - | 3 | 319 | |||
AK166645.1 | - | 3 | 281 | |||
BC024891.1 | - | 3 | 273 | 3992247 |