Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025813.3 | NP_080089.1 | 3 | 367 | ||
GenBank mRNA | AK004586.1 | - | 3 | 368 | ||
AK032688.1 | - | 3 | 336 | |||
AK075859.1 | - | 3 | 397 | |||
AK135235.1 | - | 3 | 368 | |||
AK136108.1 | - | 3 | 359 | |||
AK152444.1 | - | 3 | 389 | |||
AK159554.1 | - | 3 | 395 | |||
AK218750.1 | - | 1 | 79 | |||
BC024891.1 | - | 3 | 349 | 3992247 |