Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025825.3 | NP_080101.1 | 10, 11 | 1466 | ||
GenBank mRNA | AK004888.1 | - | 10, 11 | 1476 | ||
AK005440.1 | - | 10, 11 | 1172 | |||
AK122222.1 | - | 1073 | ||||
AK133853.1 | - | 10, 11 | 1458 | |||
AK214796.1 | - | 2, 3 | 133 | |||
BC018442.1 | - | 10, 11 | 1444 | 3157290 | ||
BC019152.1 | - | 10, 11 | 1467 | 5065139 |