Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lman2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025828.3 | NP_080104.2 | 4 | 696 | ||
GenBank mRNA | AK004952.1 | - | 4 | 527 | ||
AK014384.1 | - | 4 | 518 | |||
AK083442.1 | - | 4 | 528 | |||
AK129028.1 | - | 4 | 523 | |||
AK150383.1 | - | 4 | 525 | |||
AK166953.1 | - | 4 | 533 | |||
AK169401.1 | - | 4 | 524 | |||
BC049221.1 | - | 4 | 520 | 2599070 | ||
BC055327.1 | - | 4 | 696 | 5702182 |