Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lman2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025828.3 | NP_080104.2 | 8 | 1356 | ||
GenBank mRNA | AK004952.1 | - | 8 | 1187 | ||
AK014384.1 | - | 8 | 1178 | |||
AK083442.1 | - | 8 | 1188 | |||
AK150383.1 | - | 8 | 1185 | |||
AK166953.1 | - | 8 | 1193 | |||
AK169401.1 | - | 8 | 1184 | |||
AK215546.1 | - | 1 | 82 | |||
BC049221.1 | - | 8 | 1180 | 2599070 | ||
BC055327.1 | - | 8 | 1356 | 5702182 |