Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025829.4 | NP_080105.1 | 2, 3 | 209 | ||
GenBank mRNA | AK005054.2 | - | 2, 3 | 210 | ||
AK007268.1 | - | 2, 3 | 198 | |||
AK035794.1 | - | 236 | ||||
AY628329.1 | - | 2, 3 | 190 | |||
BC027014.1 | - | 2, 3 | 204 | 5369891 |