Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025829.4 | NP_080105.1 | 7 | 1372 | ||
GenBank mRNA | AK005054.2 | - | 7 | 1373 |