Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025832.2 | NP_080108.1 | 4, 5 | 455 | ||
GenBank mRNA | AK005056.1 | - | 4, 5 | 465 | ||
AK007755.1 | - | 4, 5 | 491 | |||
AK028432.1 | - | 4, 5 | 565 | |||
AK152498.1 | - | 4, 5 | 465 | |||
BC052445.1 | - | 4, 5 | 455 | 6412745 |