Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baiap2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025833.3 | NP_080109.1 | 9 | 1153 | ||
GenBank mRNA | AK004918.2 | - | 9 | 1154 | ||
AK008203.1 | - | 9 | 1054 | |||
AK162158.1 | - | 1183 | ||||
BC015459.1 | - | 9 | 931 | 3592632 |