Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025839.3 | NP_080115.1 | 3 | 1053 | ||
GenBank mRNA | AF435792.1 | - | 3 | 1065 | ||
AK002378.1 | - | 3 | 1046 | |||
AK003241.1 | - | 3 | 1054 | |||
AK007897.1 | - | 3 | 1049 | |||
AK159841.1 | - | 3 | 1046 | |||
BC025569.1 | - | 3 | 1127 | 5322456 |