Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025846.2 | NP_080122.2 | 6 | 1154 | ||
GenBank mRNA | AK003733.1 | - | 3 | 540 | ||
AK011419.1 | - | 6 | 1154 | |||
AK078870.1 | - | 6 | 1155 | |||
AK172037.1 | - | 6 | 1146 | |||
AK172113.1 | - | 6 | 2426 | |||
BC003871.1 | - | 6 | 1105 | 3492159 |