Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025862.2 | NP_080138.2 | 3 | 408 | ||
GenBank mRNA | AK009379.1 | - | 3 | 387 | ||
AK019502.1 | - | 3 | 405 | |||
AK029897.1 | - | 3 | 372 | |||
AK051091.1 | - | 3 | 363 | |||
AK077335.1 | - | 3 | 388 | |||
AK161255.1 | - | 3 | 362 | |||
AK178437.1 | - | 1 | 53 | |||
AK211554.1 | - | 1 | 127 | |||
BC026535.1 | - | 3 | 382 | |||
BC037644.1 | - | 3 | 384 | 5053565 | ||
BQ931973.1 | - | 3 | 408 |