Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167820.1 | NP_001161292.1 | 3 | 1206 | ||
NM_001167821.1 | NP_001161293.1 | 2 | 1036 | |||
NM_001167822.2 | NP_001161294.1 | 2 | 1093 | |||
NM_001167823.1 | NP_001161295.1 | 2 | 1109 | |||
NM_001317074.1 | NP_001304003.1 | 2 | 1086 | |||
NM_001317075.1 | NP_001304004.1 | 2 | 1002 | |||
NM_006698.3 | NP_006689.1 | 2 | 1073 | |||
XM_017027599.1 | XP_016883088.1 | 2 | 1986 | |||
GenBank mRNA | AF053470.1 | - | 2 | 1012 | ||
AK074498.1 | - | 5 | 822 | |||
AK124822.1 | - | 3 | 931 | |||
AL049288.1 | - | 2 | 1019 | |||
BC047692.1 | - | 2 | 1004 | 6059195 | ||
BC071704.1 | - | 2 | 998 | 5190373 |