Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025862.2 | NP_080138.2 | 5 | 581 | ||
GenBank mRNA | AK009379.1 | - | 5 | 560 | ||
AK019502.1 | - | 5 | 578 | |||
AK029897.1 | - | 5 | 545 | |||
AK049908.1 | - | 1 | 901 | |||
AK051091.1 | - | 5 | 536 | |||
AK077335.1 | - | 5 | 561 | |||
AK161255.1 | - | 5 | 535 | |||
AK207169.1 | - | 2 | 69 | |||
AK211554.1 | - | 3 | 301 | |||
BC026535.1 | - | 5 | 555 | |||
BC037644.1 | - | 5 | 557 | 5053565 | ||
BQ931973.1 | - | 5 | 581 |