Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025869.3 | NP_080145.1 | 5 | 1518 | ||
GenBank mRNA | AB104416.1 | - | 3 | 1310 | ||
AK007513.1 | - | 3 | 888 | |||
AK009781.1 | - | 5 | 1519 | |||
BC018204.1 | - | 3 | 1310 | 4038120 |