Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102407.1 | NP_001095877.1 | 6 | 1266 | ||
NM_025875.2 | NP_080151.2 | 6 | 1263 | |||
GenBank mRNA | AK009953.1 | - | 6 | 1257 |