Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025890.3 | NP_080166.1 | 3 | 1388 | ||
GenBank mRNA | AB211060.1 | - | 3 | 1325 | ||
AK010377.1 | - | 3 | 1325 | |||
AK021223.1 | - | 1 | 561 | |||
AK166731.1 | - | 3 | 1343 | |||
BC137808.1 | - | 3 | 1296 | 8860827 | ||
BC137809.1 | - | 3 | 1296 | 8860828 | ||
BY756270.1 | - | 1 | 400 | |||
DQ917421.1 | - | 3 | 1276 | |||
DQ917422.1 | - | 4 | 1204 |