Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025890.3 | NP_080166.1 | 3 | 1465 | ||
GenBank mRNA | AB211060.1 | - | 3 | 1402 | ||
AK010377.1 | - | 3 | 1402 | |||
AK021223.1 | - | 1 | 638 | |||
AK166731.1 | - | 3 | 1420 | |||
BC137808.1 | - | 3 | 1373 | 8860827 | ||
BC137809.1 | - | 3 | 1373 | 8860828 | ||
BY756270.1 | - | 1 | 477 |