Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025894.2 | NP_080170.1 | 11 | 1692 | ||
GenBank mRNA | AK005114.1 | - | 11 | 1726 | ||
AK008646.1 | - | 11 | 1691 | |||
AK009969.1 | - | 11 | 1695 | |||
AK151242.1 | - | 11 | 1693 | |||
AK160283.1 | - | 11 | 1715 | |||
AK167236.1 | - | 11 | 1699 |